Assay Details
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133847.3 | NP_598608.2 | 2 - 3 | 384 | 67 | |
GenBank mRNA | AB093220.2 | - | 1 - 2 | 128 | 67 | |
AK035622.1 | - | 2 - 3 | 346 | 67 | ||
AK047497.1 | - | 2 - 3 | 138 | 67 | ||
AK048158.1 | - | 2 - 3 | 142 | 67 | ||
AK083541.1 | - | 2 - 3 | 384 | 67 | ||
AK087048.1 | - | 2 - 3 | 137 | 67 | ||
AK088012.1 | - | 2 - 3 | 140 | 67 | ||
AK162435.1 | - | 2 - 3 | 145 | 67 | ||
AK171487.1 | - | 2 - 3 | 142 | 67 | ||
BC110309.1 | - | 2 - 3 | 140 | 5324218 | 67 | |
BC116320.2 | - | 2 - 3 | 380 | 40096002 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC116320.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
