Assay Details
Gene Symbol: Rfx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024918.1 | NP_001020089.1 | 11 - 12 | 1567 | 111 | |
NM_027689.3 | NP_081965.2 | 7 - 8 | 899 | 111 | ||
GenBank mRNA | AB086957.1 | - | 11 - 12 | 1421 | 111 | |
AB089184.1 | - | 7 - 8 | 900 | 111 | ||
AK016791.1 | - | 7 - 8 | 900 | 111 | ||
AK034131.1 | - | 11 - 12 | 1554 | 111 | ||
AK132673.1 | - | 8 - 9 | 962 | 111 | ||
AY102010.2 | - | 11 - 12 | 1450 | 111 | ||
AY342003.1 | - | 11 - 12 | 1432 | 111 | ||
BC138719.1 | - | 7 - 8 | 887 | 8861741 | 111 | |
BC138720.1 | - | 7 - 8 | 887 | 8861742 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
