Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168591.1 | NP_001162063.1 | 6 - 7 | 774 | 77 | |
NM_025827.3 | NP_080103.1 | 12 - 13 | 2078 | 77 | ||
GenBank mRNA | AK004843.1 | - | 12 - 13 | 1992 | 77 | |
AK050231.1 | - | 12 - 13 | 1987 | 77 | ||
AK075719.1 | - | 6 - 7 | 775 | 77 | ||
AK088348.1 | - | 11 - 12 | 1984 | 77 | ||
AK151103.1 | - | 12 - 13 | 2024 | 77 | ||
AK151406.1 | - | 10 - 11 | 1592 | 77 | ||
AK152964.1 | - | 10 - 11 | 1593 | 77 | ||
AK168628.1 | - | 10 - 11 | 1602 | 77 | ||
AK168791.1 | - | 12 - 13 | 2024 | 77 | ||
AK168956.1 | - | 12 - 13 | 2024 | 77 | ||
BC049090.1 | - | 12 - 13 | 2014 | 6401542 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
