Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025827.3 | NP_080103.1 | 1 - 2 | 371 | 106 | |
GenBank mRNA | AK004843.1 | - | 1 - 2 | 285 | 106 | |
AK050231.1 | - | 1 - 2 | 280 | 106 | ||
AK088348.1 | - | 1 - 2 | 372 | 106 | ||
AK151103.1 | - | 1 - 2 | 317 | 106 | ||
AK168628.1 | - | 1 - 2 | 321 | 106 | ||
AK168791.1 | - | 1 - 2 | 317 | 106 | ||
AK168956.1 | - | 1 - 2 | 317 | 106 | ||
AK169656.1 | - | 285 | 106 | |||
AK200906.1 | - | 1 - 2 | 83 | 106 | ||
BC049090.1 | - | 1 - 2 | 307 | 6401542 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
