Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025827.3 | NP_080103.1 | 5 - 6 | 1034 | 112 | |
GenBank mRNA | AK004843.1 | - | 5 - 6 | 948 | 112 | |
AK050231.1 | - | 5 - 6 | 943 | 112 | ||
AK151103.1 | - | 5 - 6 | 980 | 112 | ||
AK168791.1 | - | 5 - 6 | 980 | 112 | ||
AK168956.1 | - | 5 - 6 | 980 | 112 | ||
AK169656.1 | - | 948 | 112 | |||
AK218948.1 | - | 2 - 3 | 299 | 112 | ||
BC049090.1 | - | 5 - 6 | 970 | 6401542 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
