Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168591.1 | NP_001162063.1 | 3 - 4 | 371 | 70 | |
NM_025827.3 | NP_080103.1 | 9 - 10 | 1675 | 70 | ||
GenBank mRNA | AK004843.1 | - | 9 - 10 | 1589 | 70 | |
AK050231.1 | - | 9 - 10 | 1584 | 70 | ||
AK075719.1 | - | 3 - 4 | 372 | 70 | ||
AK088348.1 | - | 8 - 9 | 1581 | 70 | ||
AK151103.1 | - | 9 - 10 | 1621 | 70 | ||
AK151406.1 | - | 7 - 8 | 1189 | 70 | ||
AK152964.1 | - | 7 - 8 | 1190 | 70 | ||
AK168628.1 | - | 7 - 8 | 1199 | 70 | ||
AK168791.1 | - | 9 - 10 | 1621 | 70 | ||
AK168956.1 | - | 9 - 10 | 1621 | 70 | ||
AK169656.1 | - | 1589 | 70 | |||
BC049090.1 | - | 9 - 10 | 1611 | 6401542 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
