Assay Details
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167910.1 | NP_001161382.1 | 20 - 21 | 3138 | 84 | |
NM_019709.4 | NP_062683.3 | 21 - 22 | 3287 | 84 | ||
GenBank mRNA | AF094820.1 | - | 19 - 20 | 2706 | 84 | |
AK029048.1 | - | 20 - 21 | 3115 | 84 | ||
AK122202.1 | - | 20 - 21 | 3103 | 84 | ||
AK147336.1 | - | 19 - 20 | 5707 | 84 | ||
AK147495.1 | - | 19 - 20 | 5540 | 84 | ||
BC011533.1 | - | 20 - 21 | 3101 | 84 | ||
BC054837.1 | - | 19 - 20 | 2906 | 6490394 | 84 | |
BC057198.1 | - | 21 - 22 | 3253 | 5310414 | 84 | |
DQ913734.1 | - | 19 - 20 | 2769 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
