Assay Details
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167910.1 | NP_001161382.1 | 4 - 5 | 1066 | 82 | |
NM_019709.4 | NP_062683.3 | 5 - 6 | 1215 | 82 | ||
GenBank mRNA | AF094820.1 | - | 3 - 4 | 634 | 82 | |
AK029048.1 | - | 4 - 5 | 1043 | 82 | ||
AK122202.1 | - | 4 - 5 | 1032 | 82 | ||
AK147336.1 | - | 4 - 5 | 1011 | 82 | ||
AK147495.1 | - | 4 - 5 | 1040 | 82 | ||
BC011533.1 | - | 4 - 5 | 1032 | 82 | ||
BC054837.1 | - | 3 - 4 | 834 | 6490394 | 82 | |
BC057198.1 | - | 5 - 6 | 1181 | 5310414 | 82 | |
DQ913734.1 | - | 3 - 4 | 697 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
