Assay Details
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 2 - 3 | 565 | 74 | |
NM_001113181.1 | NP_001106652.1 | 2 - 3 | 565 | 74 | ||
NM_019691.4 | NP_062665.3 | 2 - 3 | 565 | 74 | ||
GenBank mRNA | AB022913.1 | - | 2 - 3 | 299 | 74 | |
AB079073.1 | - | 1 - 2 | 86 | 74 | ||
AB102777.1 | - | 1 - 2 | 86 | 74 | ||
AK031568.1 | - | 2 - 3 | 539 | 74 | ||
AK032103.1 | - | 2 - 3 | 566 | 74 | ||
AK047179.1 | - | 2 - 3 | 444 | 74 | ||
AK135034.1 | - | 2 - 3 | 271 | 74 | ||
BC060697.1 | - | 2 - 3 | 473 | 6409918 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK031568.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
