Assay Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284228.1 | NP_001271157.1 | 2 - 3 | 102 | 66 | |
NM_008028.2 | NP_032054.1 | 2 - 3 | 246 | 66 | ||
GenBank mRNA | BC070423.1 | - | 2 - 3 | 204 | 6403816 | 66 |
U07890.1 | - | 2 - 3 | 116 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC070423.1, NM_001284228.1, NM_008028.2, U07890.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
