Assay Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144825.2 | NP_659074.2 | 7 - 8 | 1268 | 74 | |
GenBank mRNA | AK138443.1 | - | 1240 | 74 | ||
AK143530.1 | - | 7 - 8 | 1275 | 74 | ||
BC047271.1 | - | 6 - 7 | 615 | 74 | ||
BC151012.1 | - | 7 - 8 | 1062 | 9087925 | 74 | |
BC151015.1 | - | 7 - 8 | 1062 | 9087928 | 74 | |
BC151016.1 | - | 7 - 8 | 1061 | 9087929 | 74 | |
CF164486.1 | - | 3 - 4 | 256 | 74 | ||
CN721819.1 | - | 5 - 6 | 419 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
