Assay Details
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 8 - 9 | 1165 | 84 | |
GenBank mRNA | BC031778.1 | - | 712 | 4501115 | 84 | |
BC049904.1 | - | 712 | 4501115 | 84 | ||
BC064815.1 | - | 8 - 9 | 1150 | 6516981 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
