Assay Details
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282000.1 | NP_001268929.1 | 2 - 3 | 480 | 101 | |
NM_001282001.1 | NP_001268930.1 | 2 - 3 | 480 | 101 | ||
NM_011250.4 | NP_035380.3 | 2 - 3 | 480 | 101 | ||
GenBank mRNA | AK142172.1 | - | 2 - 3 | 439 | 101 | |
AK155256.1 | - | 2 - 3 | 444 | 101 | ||
AK160027.1 | - | 2 - 3 | 427 | 101 | ||
BC020528.1 | - | 2 - 3 | 428 | 4240592 | 101 | |
BY749910.1 | - | 2 - 3 | 444 | 101 | ||
U36799.1 | - | 2 - 3 | 380 | 101 | ||
U47333.1 | - | 2 - 3 | 422 | 101 | ||
U50850.1 | - | 2 - 3 | 388 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
