Assay Details
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010764.2 | NP_034894.2 | 10 - 11 | 1364 | 63 | |
GenBank mRNA | AK004817.2 | - | 10 - 11 | 1321 | 63 | |
AK035551.1 | - | 10 - 11 | 1319 | 63 | ||
AK147928.1 | - | 10 - 11 | 1322 | 63 | ||
AK150154.1 | - | 7 - 8 | 696 | 63 | ||
AK170848.1 | - | 7 - 8 | 695 | 63 | ||
AK171166.1 | - | 10 - 11 | 1322 | 63 | ||
AK171377.1 | - | 7 - 8 | 695 | 63 | ||
BC005430.1 | - | 10 - 11 | 1309 | 3488681 | 63 | |
BC023332.1 | - | 1 - 2 | 66 | 63 | ||
U29947.1 | - | 10 - 11 | 1307 | 63 | ||
U87240.1 | - | 10 - 11 | 1298 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
