Assay Details
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028007.3 | NP_082283.2 | 11 - 12 | 1432 | 136 | |
GenBank mRNA | AK009867.1 | - | 11 - 12 | 1256 | 136 | |
AK040124.1 | - | 1 - 2 | 2297 | 136 | ||
AK132242.1 | - | 11 - 12 | 1378 | 136 | ||
AK210719.1 | - | 1 - 2 | 129 | 135 | ||
BC003977.1 | - | 11 - 12 | 1392 | 3489518 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
