Assay Details
Gene Symbol: Ern1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023913.2 | NP_076402.1 | 13 - 14 | 1796 | 94 | |
GenBank mRNA | AB031332.1 | - | 13 - 14 | 1796 | 94 | |
AK077041.1 | - | 1818 | 94 | |||
AK080709.1 | - | 7 - 8 | 1049 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
