Assay Details
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016925.3 | NP_058621.2 | 10 - 11 | 899 | 66 | |
GenBank mRNA | AF208116.1 | - | 10 - 11 | 901 | 66 | |
AF208117.1 | - | 7 - 8 | 582 | 66 | ||
AF208118.1 | - | 10 - 11 | 901 | 66 | ||
AK084990.1 | - | 9 - 10 | 1007 | 66 | ||
AK166112.1 | - | 10 - 11 | 884 | 66 | ||
BC150705.1 | - | 10 - 11 | 897 | 9087616 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
