Assay Details
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133973.2 | NP_598734.1 | 12 - 13 | 1666 | 62 | |
GenBank mRNA | AK156466.1 | - | 12 - 13 | 1658 | 62 | |
AK172465.1 | - | 12 - 13 | 1657 | 62 | ||
AK188268.1 | - | 2 - 3 | 252 | 62 | ||
AK188304.1 | - | 2 - 3 | 250 | 62 | ||
AY223866.1 | - | 12 - 13 | 1693 | 62 | ||
BC023120.1 | - | 12 - 13 | 1643 | 4161858 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
