Assay Details
Gene Symbol: Phlpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122594.2 | NP_001116066.2 | 10 - 11 | 1642 | 62 | |
GenBank mRNA | AK048260.1 | - | 10 - 11 | 1599 | 62 | |
AK048472.1 | - | 10 - 11 | 1643 | 62 | ||
BC117961.1 | - | 10 - 11 | 1642 | 40098281 | 62 | |
BC117962.1 | - | 10 - 11 | 1634 | 40098282 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
