Assay Details
Gene Symbol: Trank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164659.1 | NP_001158131.1 | 14 - 15 | 5224 | 72 | |
GenBank mRNA | AK129119.1 | - | 2 - 3 | 1290 | 72 | |
BC086653.1 | - | 2 - 3 | 250 | 30536238 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC086653.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
