Assay Details
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011072.4 | NP_035202.1 | 2 - 3 | 497 | 67 | |
GenBank mRNA | AK011066.1 | - | 2 - 3 | 482 | 67 | |
AK028117.1 | - | 2 - 3 | 448 | 67 | ||
AK150780.1 | - | 2 - 3 | 454 | 67 | ||
AK152491.1 | - | 2 - 3 | 483 | 67 | ||
AK160191.1 | - | 2 - 3 | 483 | 67 | ||
BC002080.1 | - | 2 - 3 | 470 | 3490976 | 67 | |
X14425.1 | - | 2 - 3 | 386 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
