Assay Details
Gene Symbol: Kif1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153103.2 | NP_694743.2 | 5 - 6 | 733 | 99 | |
GenBank mRNA | AB001456.1 | - | 1 - 2 | 84 | 99 | |
AB074017.1 | - | 3 - 4 | 363 | 99 | ||
AK029250.1 | - | 5 - 6 | 660 | 99 | ||
AK173007.1 | - | 4 - 5 | 440 | 99 | ||
BC029286.1 | - | 5 - 6 | 720 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
