Assay Details
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083331.2 | NP_001076800.1 | 15 - 16 | 2095 | 110 | |
NM_001276406.1 | NP_001263335.1 | 15 - 16 | 2077 | 110 | ||
NM_172394.3 | NP_765982.2 | 15 - 16 | 2062 | 110 | ||
GenBank mRNA | AJ532593.1 | - | 15 - 16 | 2071 | 110 | |
AK002643.1 | - | 15 - 16 | 2063 | 110 | ||
AK028563.1 | - | 15 - 16 | 2085 | 110 | ||
AK160619.1 | - | 15 - 16 | 2063 | 110 | ||
BC032929.1 | - | 15 - 16 | 2043 | 1379673 | 110 | |
U01135.1 | - | 3 - 4 | 284 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
