Assay Details
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026482.2 | NP_080758.1 | 18 - 19 | 3502 | 67 | |
GenBank mRNA | AK161995.1 | - | 2 - 3 | 270 | 67 | |
BC025199.1 | - | 1 - 2 | 82 | 67 | ||
BC029045.1 | - | 12 - 13 | 2142 | 67 | ||
BC030901.1 | - | 12 - 13 | 2143 | 67 | ||
BC049262.1 | - | 12 - 13 | 2142 | 67 | ||
BC057180.1 | - | 3 - 4 | 517 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
