Assay Details
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163336.1 | NP_001156808.1 | 12 - 13 | 1733 | 60 | |
NM_001163337.1 | NP_001156809.1 | 12 - 13 | 1733 | 60 | ||
NM_016745.3 | NP_058025.2 | 12 - 13 | 1733 | 60 | ||
GenBank mRNA | AK089487.1 | - | 12 - 13 | 1733 | 60 | |
AK155904.1 | - | 12 - 13 | 1734 | 60 | ||
BC017639.1 | - | 12 - 13 | 1647 | 4924519 | 60 | |
BC026147.1 | - | 12 - 13 | 1679 | 4191458 | 60 | |
U49393.1 | - | 12 - 13 | 1594 | 60 | ||
U49394.1 | - | 12 - 13 | 1594 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
