Assay Details
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009671.5 | NP_033801.4 | 7 - 8 | 919 | 145 | |
GenBank mRNA | AB011370.2 | - | 7 - 8 | 917 | 145 | |
AB098329.1 | - | 7 - 8 | 917 | 145 | ||
BC139231.1 | - | 7 - 8 | 930 | 8862253 | 145 | |
BC145583.1 | - | 7 - 8 | 930 | 9054149 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
