Assay Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011101.3 | NP_035231.2 | 11 - 12 | 1542 | 74 | |
GenBank mRNA | AK147084.1 | - | 11 - 12 | 1543 | 74 | |
BC096493.1 | - | 11 - 12 | 1336 | 30742571 | 74 | |
M25811.1 | - | 11 - 12 | 1576 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
