Assay Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011101.3 | NP_035231.2 | 7 - 8 | 1038 | 78 | |
GenBank mRNA | AK147084.1 | - | 7 - 8 | 1039 | 78 | |
BC096493.1 | - | 7 - 8 | 832 | 30742571 | 78 | |
X52684.1 | - | 8 - 9 | 979 | 78 | ||
X52685.1 | - | 8 - 9 | 979 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
