Assay Details
Gene Symbol: Cdc42ep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163346.1 | NP_001156818.1 | 1 - 2 | 129 | 107 | |
GenBank mRNA | AK075739.1 | - | 1 - 2 | 122 | 107 | |
AK165318.1 | - | 1 - 2 | 130 | 107 | ||
BC003857.1 | - | 1 - 2 | 92 | 3484427 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK075739.1, AK165318.1, BC003857.1, NM_001163346.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
