Assay Details
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173755.3 | NP_776116.2 | 11 - 12 | 1918 | 84 | |
GenBank mRNA | AK129431.1 | - | 11 - 12 | 1918 | 84 | |
BC039233.1 | - | 3 - 4 | 384 | 84 | ||
BC051108.1 | - | 2 - 3 | 263 | 84 | ||
BC059193.1 | - | 11 - 12 | 1543 | 4160667 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
