Assay Details
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173755.3 | NP_776116.2 | 2 - 3 | 491 | 60 | |
GenBank mRNA | AK129431.1 | - | 2 - 3 | 491 | 60 | |
BC059193.1 | - | 2 - 3 | 116 | 4160667 | 60 | |
U20780.1 | - | 2 - 3 | 153 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC059193.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
