Assay Details
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173755.3 | NP_776116.2 | 9 - 10 | 1649 | 89 | |
GenBank mRNA | AK129431.1 | - | 9 - 10 | 1649 | 89 | |
BC039233.1 | - | 1 - 2 | 117 | 89 | ||
BC059193.1 | - | 9 - 10 | 1274 | 4160667 | 89 | |
U20780.1 | - | 9 - 10 | 1310 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
