Assay Details
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033398.2 | NP_203971.2 | 1 - 2 | 387 | 71 | |
GenBank mRNA | AK122317.1 | - | 1 - 2 | 228 | 71 | |
CX210791.1 | - | 1 - 2 | 93 | 71 | ||
EF527404.1 | - | 1 - 2 | 387 | 71 | ||
EF527405.1 | - | 1 - 2 | 173 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
