Assay Details
Gene Symbol: Kansl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081045.1 | NP_001074514.1 | 12 - 13 | 3276 | 89 | |
GenBank mRNA | AK006970.1 | - | 4 - 5 | 387 | 89 | |
AK122484.1 | - | 12 - 13 | 3282 | 89 | ||
BC025052.1 | - | 2 - 3 | 157 | 89 | ||
BC043121.1 | - | 12 - 13 | 2825 | 6409084 | 89 | |
BC054752.1 | - | 13 - 14 | 3099 | 6834541 | 89 | |
BC079594.1 | - | 12 - 13 | 2824 | 6827379 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
