Assay Details
Gene Symbol: Kansl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081045.1 | NP_001074514.1 | 6 | 2373 | 69 | |
GenBank mRNA | AK037800.1 | - | 7 | 1939 | 69 | |
AK122484.1 | - | 6 | 2379 | 69 | ||
BC043121.1 | - | 6 | 1922 | 6409084 | 69 | |
BC053389.1 | - | 5 | 1824 | 30061629 | 69 | |
BC054752.1 | - | 7 | 2196 | 6834541 | 69 | |
BC079594.1 | - | 6 | 1921 | 6827379 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
