Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013640.3 | NP_038668.2 | 2 - 3 | 526 | 63 | |
GenBank mRNA | AK144096.1 | - | 2 - 3 | 249 | 63 | |
BC004730.1 | - | 2 - 3 | 252 | 3583052 | 63 | |
CT010274.1 | - | 2 - 3 | 144 | 63 | ||
D85561.1 | - | 2 - 3 | 221 | 63 | ||
U77784.1 | - | 2 - 3 | 213 | 63 | ||
Y10875.1 | - | 2 - 3 | 244 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
