Assay Details
Gene Symbol: Trpc4ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163452.1 | NP_001156924.1 | 4 - 5 | 553 | 80 | |
NM_019828.2 | NP_062802.2 | 4 - 5 | 553 | 80 | ||
GenBank mRNA | AF509881.1 | - | 4 - 5 | 475 | 80 | |
AK076496.1 | - | 4 - 5 | 552 | 80 | ||
AK140799.1 | - | 3 - 4 | 241 | 80 | ||
AK154263.1 | - | 4 - 5 | 502 | 80 | ||
AK156047.1 | - | 4 - 5 | 502 | 80 | ||
AK165259.1 | - | 4 - 5 | 502 | 80 | ||
AK170085.1 | - | 4 - 5 | 491 | 80 | ||
AK171404.1 | - | 4 - 5 | 502 | 80 | ||
AK220239.1 | - | 4 - 5 | 529 | 80 | ||
BC033274.1 | - | 4 - 5 | 481 | 3673725 | 80 | |
BC057330.1 | - | 4 - 5 | 511 | 6816155 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
