Assay Details
Gene Symbol: Mast1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019945.2 | NP_064329.2 | 20 - 21 | 2646 | 83 | |
GenBank mRNA | AF077818.1 | - | 20 - 21 | 2629 | 83 | |
AK122411.1 | - | 10 - 11 | 1781 | 83 | ||
BC054524.1 | - | 20 - 21 | 2646 | 6400705 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
