Assay Details
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164559.1 | NP_001158031.1 | 6 - 7 | 876 | 110 | |
NM_001164560.1 | NP_001158032.1 | 6 - 7 | 800 | 110 | ||
NM_198020.2 | NP_932137.2 | 6 - 7 | 903 | 110 | ||
GenBank mRNA | AK032945.1 | - | 5 - 6 | 1049 | 110 | |
AK078395.1 | - | 5 - 6 | 1079 | 110 | ||
AK084775.1 | - | 6 - 7 | 841 | 110 | ||
AK155251.1 | - | 6 - 7 | 869 | 111 | ||
AK159467.1 | - | 6 - 7 | 841 | 110 | ||
AK170412.1 | - | 6 - 7 | 765 | 110 | ||
AK171002.1 | - | 6 - 7 | 868 | 110 | ||
BC027404.1 | - | 6 - 7 | 872 | 4950980 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
