Assay Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 8 - 9 | 1852 | 74 | |
NM_018823.2 | NP_061293.2 | 9 - 10 | 1906 | 74 | ||
NM_133957.3 | NP_598718.2 | 10 - 11 | 1968 | 74 | ||
GenBank mRNA | AF162853.1 | - | 8 - 9 | 1763 | 74 | |
AF200687.1 | - | 9 - 10 | 1541 | 74 | ||
AF228706.1 | - | 6 - 7 | 1606 | 74 | ||
AF369980.1 | - | 10 - 11 | 1607 | 74 | ||
AF453571.2 | - | 10 - 11 | 1576 | 74 | ||
AK134022.1 | - | 10 - 11 | 1972 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
