Assay Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 9 - 10 | 1990 | 132 | |
NM_018823.2 | NP_061293.2 | 10 - 11 | 2044 | 132 | ||
NM_133957.3 | NP_598718.2 | 11 - 12 | 2106 | 132 | ||
GenBank mRNA | AF162853.1 | - | 9 - 10 | 1901 | 132 | |
AF200687.1 | - | 10 - 11 | 1679 | 132 | ||
AF228706.1 | - | 7 - 8 | 1744 | 132 | ||
AF369980.1 | - | 11 - 12 | 1745 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
