Assay Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 3 - 4 | 1111 | 76 | |
NM_018823.2 | NP_061293.2 | 4 - 5 | 1165 | 76 | ||
NM_133957.3 | NP_598718.2 | 5 - 6 | 1227 | 76 | ||
GenBank mRNA | AF162853.1 | - | 3 - 4 | 1022 | 76 | |
AF200687.1 | - | 4 - 5 | 800 | 76 | ||
AF228705.1 | - | 1 - 2 | 865 | 76 | ||
AF228706.1 | - | 1 - 2 | 865 | 76 | ||
AF369980.1 | - | 5 - 6 | 866 | 76 | ||
AK134022.1 | - | 5 - 6 | 1231 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
