Assay Details
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172283.3 | NP_758487.2 | 13 - 14 | 1430 | 113 | |
GenBank mRNA | AJ297482.4 | - | 13 - 14 | 1401 | 113 | |
AJ534942.1 | - | 13 - 14 | 1401 | 113 | ||
AK049153.1 | - | 2 - 3 | 259 | 113 | ||
AY223865.1 | - | 13 - 14 | 1401 | 113 | ||
BC037698.1 | - | 12 - 13 | 1379 | 4190449 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
