Assay Details
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172283.3 | NP_758487.2 | 1 - 2 | 68 | 65 | |
NR_052009.1 | 1 - 2 | 68 | 65 | |||
GenBank mRNA | AJ297482.4 | - | 1 - 2 | 39 | 65 | |
AJ534942.1 | - | 1 - 2 | 39 | 65 | ||
AK164340.1 | - | 1 - 2 | 37 | 65 | ||
AY223865.1 | - | 1 - 2 | 39 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AJ297482.4, AJ534942.1, AY223865.1, NM_172283.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
