Assay Details
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172760.3 | NP_766348.3 | 9 - 10 | 951 | 109 | |
GenBank mRNA | AK037067.1 | - | 9 - 10 | 944 | 109 | |
AK157875.1 | - | 889 | 109 | |||
BC018516.1 | - | 7 - 8 | 840 | 3155913 | 109 | |
BC026617.1 | - | 4 - 5 | 896 | 109 | ||
BC058752.1 | - | 9 - 10 | 934 | 6485393 | 109 | |
BC138887.1 | - | 9 - 10 | 908 | 8861909 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
