Assay Details
Gene Symbol: E2f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148952.1 | NP_683754.1 | 6 - 7 | 890 | 71 | |
GenBank mRNA | AK145950.1 | - | 6 - 7 | 837 | 71 | |
AK157028.1 | - | 6 - 7 | 837 | 71 | ||
AK194813.1 | - | 2 - 3 | 342 | 71 | ||
BC023859.1 | - | 6 - 7 | 839 | 5346688 | 71 | |
BC026649.1 | - | 6 - 7 | 890 | 4987691 | 71 | |
BC027048.1 | - | 6 - 7 | 850 | 5012650 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
