Assay Details
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB089793.1 | - | 6 - 7 | 554 | 81 | |
AF482993.1 | - | 5 - 6 | 518 | 81 | ||
AK049410.1 | - | 6 - 7 | 550 | 81 | ||
AK051791.1 | - | 5 - 6 | 469 | 81 | ||
AK143281.1 | - | 6 - 7 | 523 | 81 | ||
AK173069.1 | - | 6 - 7 | 890 | 81 | ||
BC030879.1 | - | 6 - 7 | 553 | 4241988 | 81 | |
BC058947.1 | - | 6 - 7 | 524 | 5702781 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
