Assay Details
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB089793.1 | - | 8 - 9 | 736 | 84 | |
AF482993.1 | - | 7 - 8 | 700 | 84 | ||
AK049410.1 | - | 8 - 9 | 732 | 84 | ||
AK051791.1 | - | 7 - 8 | 651 | 84 | ||
AK160826.1 | - | 6 - 7 | 574 | 84 | ||
AK173069.1 | - | 8 - 9 | 1072 | 84 | ||
AK216559.1 | - | 1 - 2 | 104 | 84 | ||
BC030879.1 | - | 8 - 9 | 735 | 4241988 | 84 | |
BC034508.1 | - | 1 - 2 | 136 | 84 | ||
BC058947.1 | - | 8 - 9 | 706 | 5702781 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
