Assay Details
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277188.1 | NP_001264117.1 | 18 - 19 | 2239 | 85 | |
NR_102339.1 | 17 - 18 | 2196 | 85 | |||
GenBank mRNA | BC065114.1 | - | 17 - 18 | 2196 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
