Assay Details
Gene Symbol: Hexim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138753.2 | NP_620092.1 | 1 | 1874 | 105 | |
GenBank mRNA | AK134535.1 | - | 1 | 1874 | 105 | |
AY090614.1 | - | 1 | 1871 | 105 | ||
BC022111.1 | - | 1 | 5067620 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of signal transduction by p53 class mediator
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
snRNA binding
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of signal transduction by p53 class mediator
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
snRNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083424 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083439 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083425 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083427 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083428 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083437 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083441 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083435 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083434 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083436 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083440 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083438 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820191 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510882 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124095 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P348397 | AY090614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P348397 | AY090614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083401 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083407 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28083442 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124095 | - | - |
Whole Mouse Genome G4122A | A_52_P85403 | BC022111.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P348397 | AY090614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P348397 | NM_138753.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122059 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124095 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09096494 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09096485 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09096496 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09356428 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09356414 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09096488 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09356415 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083440 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083435 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083436 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083427 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083425 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083438 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083437 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083439 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083424 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083434 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083428 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28083441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28083441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09340942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09340948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09458818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09134689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09356423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09356425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09320879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09096485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28083447 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10381701 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381698 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381700 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381699 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419359_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425937_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10381697 | - | - |
Mouse Genome 430 2.0 Array | 1425937_a_at | - | - |
Mouse Genome 430 2.0 Array | 1425937_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1419359_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419359_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192231_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192231_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100016124.mm.1 | TC1100001706.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100016124.mm.1 | NM_138753.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100016124.mm.1 | NM_138753.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100016125.mm.1 | TC1100001706.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100016125.mm.1 | NM_138753.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100016125.mm.1 | NM_138753.2 | Refseq |
Mouse Expression Set 430 | 1419359_PM_at | - | - |
Mouse Expression Set 430 | 1425937_PM_a_at | - | - |
Mouse Expression Set 430 | 1419359_at | - | - |
Mouse Expression Set 430 | 1425937_a_at | - | - |
Murine Genome U74v2 Set | 141037_at | - | - |
Murine Genome U74v2 Set | 141037_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419359_at | - | - |
Mouse Genome 430A 2.0 Array | 1419359_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425937_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1425937_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419359_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425937_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419359_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425937_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 389131 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 7100717 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007100717 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631994 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2631994 | - | - |
mouse-6 v1.1 (Illumina) | scl40848.1_563-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7100717 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl40848.1_563-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100717 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2631994 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631994 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40848.1_563-S | - | - |
